Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smoc1
Entrez Gene ID
64075
Gene Name
SPARC related modular calcium binding 1
Gene Aliases
2600002F22Rik, AI848508, SRG
Chromosome Location
Chr.12: 81026808 - 81186414 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146217.1 NP_001139689.1 6 - 7 865 134
NM_022316.2 NP_071711.2 6 - 7 832 134
GenBank mRNA AF070470.1 - 6 - 7 808 134
AK040931.1 - 6 - 7 866 134
BC031804.1 - 6 - 7 812 134

View Full Product Details