Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdc37l1
Entrez Gene ID
67072
Gene Name
cell division cycle 37-like 1
Gene Aliases
2700033A15Rik, BB159850, Cdc37l, Harc
Chromosome Location
Chr.19: 28990325 - 29026682 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK012312.1 - 6 - 7 951 93
BC031761.1 - 6 - 7 944 93

View Full Product Details