Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cuzd1
Entrez Gene ID
16433
Gene Name
CUB and zona pellucida-like domains 1
Gene Aliases
Erg-1, Itmap1, Uo-44, Usg, Utczp
Chromosome Location
Chr.7: 131308554 - 131322292 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008411.3 NP_032437.3 8 - 9 1899 73
GenBank mRNA AK019038.1 - 1 - 2 73 73
BC140992.1 - 8 - 9 1834 73
BY707920.1 - 1 - 2 73 73
U69699.2 - 8 - 9 1898 73

View Full Product Details