Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif4
Entrez Gene ID
16571
Gene Name
kinesin family member 4
Gene Aliases
AI323435, D330050K22Rik, Kns4
Chromosome Location
Chr.X: 100626065 - 100727271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008446.2 NP_032472.2 24 - 25 3302 88
GenBank mRNA AK135430.1 - 24 - 25 2796 88
AK136346.1 - 5 - 6 543 88
AK171938.1 - 24 - 25 3214 88
BC050946.1 - 24 - 25 3240 88
BC054537.1 - 24 - 25 3224 88
D12646.1 - 24 - 25 3629 88

View Full Product Details