Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ints6
Entrez Gene ID
18130
Gene Name
integrator complex subunit 6
Gene Aliases
2900075H24Rik, AI480962, DICE1, Ddx26, HDB, Notch2l
Chromosome Location
Chr.14: 62676325 - 62761370 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008715.2 NP_032741.2 17 - 18 3009 135
GenBank mRNA AK046055.1 - 10 - 11 1594 135
BC059263.1 - 17 - 18 3009 135

View Full Product Details