The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009421.3 | NP_033447.2 | 4 - 5 | 450 | 98 |
GenBank mRNA | AK029135.1 | - | 4 - 5 | 452 | 98 |
AK089281.1 | - | 3 - 4 | 194 | 98 | |
AK142931.1 | - | 3 - 4 | 209 | 98 | |
AK169515.1 | - | 4 - 5 | 243 | 98 | |
L35302.1 | - | 3 - 4 | 196 | 98 |