The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009421.3 | NP_033447.2 | 8 - 9 | 1160 | 78 |
GenBank mRNA | AK029135.1 | - | 8 - 9 | 1162 | 78 |
AK169515.1 | - | 8 - 9 | 953 | 78 | |
AK172598.1 | - | 4 - 5 | 764 | 78 | |
BC096595.1 | - | 4 - 5 | 78 | ||
L35302.1 | - | 7 - 8 | 906 | 78 |