Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wee1
Entrez Gene ID
22390
Gene Name
WEE 1 homolog 1 (S. pombe)
Gene Aliases
Wee1A
Chromosome Location
Chr.7: 110121700 - 110143299 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009516.3 NP_033542.2 9 - 10 1925 76
GenBank mRNA AK011212.1 - 9 - 10 1701 76
AK039572.1 - 9 - 10 1241 76
BC006852.1 - 9 - 10 1944 76
D30743.1 - 9 - 10 1926 76

View Full Product Details