The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083188.1 | NP_001076657.1 | 21 - 22 | 2267 | 92 |
NM_001199310.1 | NP_001186239.1 | 21 - 22 | 2275 | 92 | |
NM_010715.2 | NP_034845.2 | 21 - 22 | 2192 | 92 | |
GenBank mRNA | AK153993.1 | - | 21 - 22 | 2154 | 92 |
AK160208.1 | - | 6 - 7 | 561 | 92 | |
AK161793.1 | - | 14 - 15 | 1350 | 92 | |
AK168643.1 | - | 21 - 22 | 2271 | 92 | |
BC028287.1 | - | 21 - 22 | 2117 | 92 | |
BC138541.1 | - | 21 - 22 | 2275 | 92 | |
BC138542.1 | - | 21 - 22 | 2275 | 92 | |
U04674.1 | - | 20 - 21 | 2060 | 92 | |
U19604.1 | - | 21 - 22 | 2305 | 92 |