The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011665.4 | NP_035795.1 | 1 - 2 | 142 | 95 |
GenBank mRNA | AK005058.1 | - | 1 - 2 | 124 | 95 |
AK088508.1 | - | 1 - 2 | 127 | 95 | |
U94402.1 | - | 1 - 2 | 138 | 95 |