The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011717.4 | NP_035847.2 | 3 - 4 | 917 | 61 |
NM_212438.3 | NP_997603.3 | 2 - 3 | 570 | 61 | |
GenBank mRNA | AB012265.1 | - | 7 - 8 | 3102 | 61 |
AB012266.1 | - | 3 - 4 | 611 | 61 | |
AB255388.1 | - | 2 - 3 | 512 | 61 | |
CA327606.1 | - | 2 - 3 | 485 | 61 | |
CB249930.1 | - | 2 - 3 | 556 | 61 |