Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd1
Entrez Gene ID
107765
Gene Name
ankyrin repeat domain 1 (cardiac muscle)
Gene Aliases
Alrp, CARP, Crap, MARP1
Chromosome Location
Chr.19: 36111965 - 36119844 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013468.3 NP_038496.2 5 - 6 612 88
GenBank mRNA AF041847.1 - 5 - 6 563 88
AK009655.1 - 5 - 6 605 88
AK009959.1 - 5 - 6 611 88
AK145944.1 - 5 - 6 653 88
AK146471.1 - 5 - 6 609 88
AK146627.1 - 5 - 6 611 88
AK146940.1 - 5 - 6 609 88
BC037138.1 - 5 - 6 612 88

View Full Product Details