Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shroom3
Entrez Gene ID
27428
Gene Name
shroom family member 3
Gene Aliases
AL022960, D5Ertd287e, Shrm, Shrm3
Chromosome Location
Chr.5: 92683435 - 92965759 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077595.2 NP_001071063.1 9 - 10 5429 75
NM_001077596.2 NP_001071064.1 9 - 10 5477 75
NM_015756.2 NP_056571.2 10 - 11 5801 75
GenBank mRNA AF199421.1 - 10 - 11 5819 75
AF199422.1 - 9 - 10 5387 75
AK049347.1 - 6 - 7 3335 75
AK129371.1 - 6 - 7 4807 75
AK147493.1 - 9 - 10 5412 75
AK164918.1 - 9 - 10 5388 75
BC003909.1 - 4 - 5 1073 75
BC145113.1 - 11 - 12 5218 75

View Full Product Details