The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016886.4 | NP_058582.3 | 15 - 16 | 2933 | 95 |
NR_104053.1 | 16 - 17 | 3048 | 95 | ||
GenBank mRNA | AB022342.1 | - | 15 - 16 | 2723 | 95 |
AK135043.1 | - | 15 - 16 | 2687 | 95 | |
AK138615.1 | - | 14 - 15 | 3394 | 95 | |
BC076584.1 | - | 16 - 17 | 2910 | 95 | |
BC086678.1 | - | 15 - 16 | 2878 | 95 | |
BC129855.1 | - | 15 - 16 | 2723 | 95 | |
BC129856.1 | - | 15 - 16 | 2723 | 95 |