Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myot
Entrez Gene ID
58916
Gene Name
myotilin
Gene Aliases
5530402I04Rik, Ttid
Chromosome Location
Chr.18: 44334053 - 44355740 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033621.3 NP_001028793.1 7 - 8 1291 119
GenBank mRNA AF230979.2 - 7 - 8 1290 119
AK017447.1 - 7 - 8 1292 119
BC016214.1 - 7 - 8 1269 119
BC064052.1 - 7 - 8 1264 119

View Full Product Details