The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022417.3 | NP_071862.2 | 2 - 3 | 424 | 70 |
GenBank mRNA | AB030199.1 | - | 2 - 3 | 367 | 70 |
AF272044.1 | - | 2 - 3 | 331 | 70 | |
AF282981.1 | - | 2 - 3 | 352 | 70 | |
AK036377.1 | - | 2 - 3 | 370 | 70 | |
AK156930.1 | - | 2 - 3 | 327 | 70 | |
AK210599.1 | - | 1 - 2 | 97 | 70 | |
AK213826.1 | - | 1 - 2 | 97 | 70 | |
AK216877.1 | - | 1 - 2 | 97 | 70 | |
BC012952.1 | - | 2 - 3 | 302 | 70 |