Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Irx3
Entrez Gene ID
16373
Gene Name
Iroquois related homeobox 3
Gene Aliases
AI894186
Chromosome Location
Chr.8: 91798511 - 91801654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008393.3 NP_032419.2 2 - 3 1816 98
GenBank mRNA BC067195.1 - 2 - 3 1815 98
BC085500.1 - 2 - 3 1536 98
Y15001.1 - 2 - 3 1402 98

View Full Product Details