Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Per1
Entrez Gene ID
18626
Gene Name
period circadian clock 1
Gene Aliases
Per, m-rigui, mPer1
Chromosome Location
Chr.11: 69095140 - 69109965 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159367.1 NP_001152839.1 21 - 22 3618 69
NM_011065.4 NP_035195.2 21 - 22 3633 69
GenBank mRNA AB002108.1 - 20 - 21 3447 69
AF022992.1 - 21 - 22 3672 69
AK148202.1 - 21 - 22 3625 69
AK172958.1 - 21 - 22 3618 69
BC039768.1 - 20 - 21 3518 69
BC091645.1 - 21 - 22 3302 69

View Full Product Details