Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spta1
Entrez Gene ID
20739
Gene Name
spectrin alpha, erythrocytic 1
Gene Aliases
AF093576, AI451697, Spna-1, Spna1, ha, ihj, nmf4, sph
Chromosome Location
Chr.1: 174172776 - 174248449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011465.4 NP_035595.2 39 - 40 5720 94
GenBank mRNA AF093576.1 - 39 - 40 5628 94
AK143823.1 - 3 - 4 388 94
AK161449.1 - 10 - 11 1361 94
BC150747.1 - 39 - 40 5585 94
U87455.1 - 39 - 40 5700 94

View Full Product Details