Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp2r3c
Entrez Gene ID
59032
Gene Name
protein phosphatase 2, regulatory subunit B'', gamma
Gene Aliases
4930511A21Rik, 5730412A08Rik, G4-1, G5pr
Chromosome Location
Chr.12: 55280809 - 55303005 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021529.3 NP_067504.2 7 - 8 802 61
GenBank mRNA AB041577.1 - 7 - 8 791 61
AJ238247.2 - 6 - 7 629 61
AK013403.1 - 6 - 7 598 61
AK015746.1 - 6 - 7 631 61
AK028063.1 - 7 - 8 767 61
AK028254.1 - 6 - 7 721 61
AK033405.1 - 7 - 8 801 61
AK040589.1 - 6 - 7 630 61
AK087572.1 - 6 - 7 625 61
AK088240.1 - 6 - 7 711 61
AK163192.1 - - 801 61
AK168298.1 - 7 - 8 804 61
BC024754.1 - 7 - 8 765 61
BC138295.1 - 6 - 7 598 61
BC138296.1 - 6 - 7 598 61

View Full Product Details