Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gtf2a1l
- Entrez Gene ID
-
71828
- Gene Name
- general transcription factor IIA, 1-like
- Gene Aliases
-
1700011N16Rik, Alf, Gtf2a1lf
- Chromosome Location
-
Chr.17: 88665502 - 88715157 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF250835.1 |
- |
1 - 2 |
41 |
70 |
View Full Product Details