Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jkamp
Entrez Gene ID
104771
Gene Name
JNK1/MAPK8-associated membrane protein
Gene Aliases
1200003C05Rik, 2310047L11Rik, Jamp
Chromosome Location
Chr.12: 72085839 - 72101527 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024205.2 NP_077167.1 1 - 2 347 73
GenBank mRNA BC002238.1 - 1 - 2 63 73

View Full Product Details