The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163316.1 | NP_001156788.1 | 2 - 3 | 184 | 84 |
NM_026269.2 | NP_080545.2 | 2 - 3 | 375 | 84 | |
GenBank mRNA | AK003551.1 | - | 2 - 3 | 324 | 84 |
BC019557.1 | - | 2 - 3 | 166 | 84 | |
CJ053487.1 | - | 2 - 3 | 376 | 84 |