Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep57l1
Entrez Gene ID
103268
Gene Name
centrosomal protein 57-like 1
Gene Aliases
2410017P07Rik, 4930484D11Rik, AI607327, cep57R
Chromosome Location
Chr.10: 41718840 - 41809868 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243074.1 NP_001230003.1 6 - 7 742 112
NM_001243075.1 NP_001230004.1 4 - 5 433 112
NM_026643.5 NP_080919.3 6 - 7 660 112
NM_029132.2 NP_083408.1 6 - 7 728 112
GenBank mRNA AK010530.1 - 6 - 7 679 112
AK012710.1 - 4 - 5 379 112
AK015619.1 - 5 - 6 643 112
BC021375.1 - 6 - 7 752 112
BC138230.1 - 6 - 7 663 112
CD354087.1 - 3 - 4 410 112

View Full Product Details