Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ints12
Entrez Gene ID
71793
Gene Name
integrator complex subunit 12
Gene Aliases
1110020M19Rik, 2810027J24Rik, 4930529N21Rik, A230056J18Rik, AI666778, Phf22
Chromosome Location
Chr.3: 133091953 - 133110988 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027927.3 NP_082203.1 2 - 3 203 101
GenBank mRNA AK003874.1 - 2 - 3 215 101
AK155162.1 - 2 - 3 215 101
BC023801.1 - 2 - 3 203 101
BC046996.1 - 2 - 3 197 101

View Full Product Details