Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ints12
Entrez Gene ID
71793
Gene Name
integrator complex subunit 12
Gene Aliases
1110020M19Rik, 2810027J24Rik, 4930529N21Rik, A230056J18Rik, AI666778, Phf22
Chromosome Location
Chr.3: 133091953 - 133110988 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027927.3 NP_082203.1 4 - 5 532 89
GenBank mRNA AK003874.1 - 4 - 5 544 89
AK012829.1 - 4 - 5 544 89
AK155162.1 - 4 - 5 544 89
BC010657.1 - 2 - 3 204 89
BC023801.1 - 4 - 5 532 89
BC046996.1 - 4 - 5 526 89

View Full Product Details