Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sncb
Entrez Gene ID
104069
Gene Name
synuclein, beta
Gene Aliases
AI838531, betaSYN
Chromosome Location
Chr.13: 54758860 - 54766440 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033610.2 NP_291088.1 1 - 2 131 70
GenBank mRNA BC019409.1 - 1 - 2 131 70

View Full Product Details