Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Helb
Entrez Gene ID
117599
Gene Name
helicase (DNA) B
Gene Aliases
AI447783, D10Ertd664e
Chromosome Location
Chr.10: 120083608 - 120112965 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080446.2 NP_536694.2 8 - 9 2285 99
GenBank mRNA AB048542.1 - 8 - 9 2210 99
AK131122.1 - 8 - 9 2307 99
BC068140.1 - 8 - 9 2213 99

View Full Product Details