The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133242.3 | NP_573505.2 | 13 - 14 | 1590 | 110 |
GenBank mRNA | AK147076.1 | - | 13 - 14 | 1535 | 110 |
BC004000.1 | - | 9 - 10 | 922 | 110 | |
BC023205.1 | - | 13 - 14 | 1492 | 110 | |
BC086645.1 | - | 13 - 14 | 1513 | 110 |