The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133945.1 | NP_598706.1 | 14 - 15 | 1461 | 79 |
GenBank mRNA | AK150435.1 | - | 14 - 15 | 1552 | 79 |
AK171673.1 | - | 14 - 15 | 1466 | 79 | |
BC010473.1 | - | 13 - 14 | 1444 | 79 | |
BC024782.1 | - | 13 - 14 | 1444 | 79 | |
BC024839.1 | - | 14 - 15 | 1461 | 79 | |
BC034196.1 | - | 13 - 14 | 1442 | 79 | |
S71575.1 | - | 6 - 7 | 696 | 79 |