Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddx1
Entrez Gene ID
104721
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 1
Gene Aliases
AA409185, DBP-RB
Chromosome Location
Chr.12: 13219307 - 13249174 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134040.1 NP_598801.1 16 - 17 1253 65
GenBank mRNA AK028341.1 - 16 - 17 1250 65
AK132157.1 - 16 - 17 1293 65
AK153335.1 - 16 - 17 1277 65
AK160982.1 - 16 - 17 1254 65
AK166146.1 - 16 - 17 1236 65
BC008570.1 - 16 - 17 1231 65
BC010624.1 - 16 - 17 1253 65

View Full Product Details