Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc17a3
Entrez Gene ID
105355
Gene Name
solute carrier family 17 (sodium phosphate), member 3
Gene Aliases
AW261723, Npt4
Chromosome Location
Chr.13: 23836918 - 23860718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164743.1 NP_001158215.1 4 - 5 763 70
NM_134069.3 NP_598830.2 5 - 6 887 70
GenBank mRNA AK128896.1 - 5 - 6 871 70
AK143896.1 - 5 - 6 859 70
BC018252.1 - 5 - 6 816 70
BC024459.1 - 5 - 6 843 70
BC034179.1 - 5 - 6 707 70
BC034183.1 - 5 - 6 692 70

View Full Product Details