The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134251.2 | NP_599012.2 | 8 - 9 | 1057 | 57 |
GenBank mRNA | AF319951.1 | - | 8 - 9 | 1051 | 57 |
AK087781.1 | - | 4 - 5 | 646 | 57 |