Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bnipl
Entrez Gene ID
171388
Gene Name
BCL2/adenovirus E1B 19kD interacting protein like
Gene Aliases
1700128A13Rik, BNIP-S, BNIPL1, BNIPL2, PP73, PP753
Chromosome Location
Chr.3: 95241293 - 95251193 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168356.1 NP_001161828.1 2 - 3 291 71
NM_134253.2 NP_599014.2 3 - 4 387 71
GenBank mRNA AK142472.1 - 2 - 3 148 71
BC005659.1 - 3 - 4 296 71

View Full Product Details