The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009659.2 | NP_033789.1 | 1 - 2 | 323 | 60 |
GenBank mRNA | AF059251.1 | - | 1 - 2 | 201 | 60 |
BC113149.1 | - | 1 - 2 | 287 | 60 | |
Y14334.1 | - | 1 - 2 | 323 | 60 |