The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020033.1 | NP_064417.1 | 4 - 5 | 459 | 65 |
GenBank mRNA | AB076256.1 | - | 4 - 5 | 471 | 65 |
AJ011118.1 | - | 4 - 5 | 459 | 65 | |
AJ245514.1 | - | 4 - 5 | 459 | 65 | |
AK132585.1 | - | 4 - 5 | 469 | 65 | |
BC138760.1 | - | 4 - 5 | 497 | 65 | |
BC138761.1 | - | 4 - 5 | 497 | 65 |