The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276442.1 | NP_001263371.1 | 12 - 13 | 1035 | 86 |
NM_025371.3 | NP_079647.1 | 12 - 13 | 1039 | 86 | |
GenBank mRNA | AK002423.1 | - | 12 - 13 | 1025 | 86 |
AK003703.1 | - | 12 - 13 | 1040 | 86 | |
BC005631.1 | - | 12 - 13 | 1043 | 86 |