The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025473.3 | NP_079749.1 | 8 - 9 | 1043 | 65 |
GenBank mRNA | AK007715.1 | - | 8 - 9 | 1044 | 65 |
AK012353.1 | - | 4 - 5 | 371 | 65 | |
BC011237.1 | - | 8 - 9 | 1041 | 65 | |
BC019752.1 | - | 9 - 10 | 809 | 65 | |
BC054411.1 | - | 8 - 9 | 716 | 65 | |
BC069871.1 | - | 8 - 9 | 1178 | 65 |