The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164082.1 | NP_001157554.1 | 6 - 7 | 1032 | 80 |
NM_025945.3 | NP_080221.3 | 7 - 8 | 1064 | 80 | |
GenBank mRNA | AK013165.1 | - | 6 - 7 | 949 | 80 |
AK156587.1 | - | 7 - 8 | 1016 | 80 | |
BC016102.1 | - | 7 - 8 | 1040 | 80 |