Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhx40
Entrez Gene ID
67487
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 40
Gene Aliases
2410016C14Rik, ARG147, DDX40, PAD
Chromosome Location
Chr.11: 86768849 - 86807660 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026191.2 NP_080467.3 6 - 7 955 72
GenBank mRNA AK004610.1 - 6 - 7 919 72
AK010512.1 - 6 - 7 935 72
AK029935.1 - 6 - 7 926 72
AK031000.1 - 6 - 7 901 72
AK135592.1 - 6 - 7 891 72
AK216252.1 - 3 - 4 341 72
BC058276.1 - 6 - 7 873 72

View Full Product Details