Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Lrp2bp
- Entrez Gene ID
-
67620
- Gene Name
- Lrp2 binding protein
- Gene Aliases
-
1700113N17Rik, 2310006J04Rik, 4930479L12Rik, MegBP
- Chromosome Location
-
Chr.8: 46010598 - 46029478 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016626.1 |
- |
- |
197 |
70 |
View Full Product Details