The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026480.3 | NP_080756.1 | 2 - 3 | 423 | 74 |
GenBank mRNA | AB283026.1 | - | 2 - 3 | 363 | 74 |
AK010266.1 | - | 2 - 3 | 418 | 74 | |
AK010809.1 | - | 2 - 3 | 424 | 74 | |
AK133554.1 | - | 2 - 3 | 388 | 74 | |
AK135950.1 | - | 2 - 3 | 389 | 74 | |
AK136199.1 | - | - | 389 | 74 | |
AK145510.1 | - | 2 - 3 | 389 | 74 | |
BC062245.1 | - | 2 - 3 | 374 | 74 | |
DQ238107.1 | - | 2 - 3 | 424 | 74 |