The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026483.2 | NP_080759.2 | 5 - 6 | 1281 | 63 |
GenBank mRNA | AK008785.1 | - | 2 - 3 | 211 | 63 |
AK013337.1 | - | 5 - 6 | 1269 | 63 | |
BC018177.1 | - | 5 - 6 | 1275 | 63 | |
BC049270.1 | - | 5 - 6 | 1257 | 63 |