Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubl7
Entrez Gene ID
69459
Gene Name
ubiquitin-like 7 (bone marrow stromal cell-derived)
Gene Aliases
2300004C15Rik
Chromosome Location
Chr.9: 57910980 - 57929970 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122873.1 NP_001116345.1 6 - 7 633 77
NM_027086.3 NP_081362.2 6 - 7 581 77
GenBank mRNA AK009025.1 - 6 - 7 582 77
AK154496.1 - 6 - 7 591 77
AK183337.1 - 2 - 3 145 77
BC016456.1 - 5 - 6 567 77
BC094047.1 - 5 - 6 570 77

View Full Product Details