Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbx
Entrez Gene ID
70508
Gene Name
bobby sox homolog (Drosophila)
Gene Aliases
5530401J07Rik, 5730403O13Rik
Chromosome Location
Chr.16: 50191844 - 50432461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027444.3 NP_081720.2 15 - 16 2746 65
GenBank mRNA AF454943.1 - 15 - 16 2739 65
AF454944.1 - 13 - 14 2611 65
AK029747.1 - 15 - 16 2800 65
AK041632.1 - 3 - 4 374 65
AK211657.1 - 3 - 4 329 65
BC066821.1 - 14 - 15 2463 65

View Full Product Details