The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028030.3 | NP_082306.2 | 7 - 8 | 880 | 64 |
GenBank mRNA | AK010301.1 | - | 8 - 9 | 718 | 64 |
AK144659.1 | - | 7 - 8 | 905 | 64 | |
AK156633.1 | - | 7 - 8 | 880 | 64 | |
BC021788.1 | - | 7 - 8 | 878 | 64 |