The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028765.3 | NP_083041.1 | 17 - 18 | 1916 | 91 |
GenBank mRNA | AK004775.1 | - | 17 - 18 | 1918 | 91 |
BC048717.1 | - | 10 - 11 | 1012 | 91 |