Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35f5
Entrez Gene ID
74150
Gene Name
solute carrier family 35, member F5
Gene Aliases
1300003P13Rik, AI646727
Chromosome Location
Chr.1: 125560414 - 125595780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028787.4 NP_083063.2 6 - 7 791 79
GenBank mRNA AK004892.2 - 6 - 7 772 79
AK076339.1 - 6 - 7 785 79
AK145848.1 - 6 - 7 792 79
AK169012.1 - 6 - 7 653 79
AK171951.1 - 6 - 7 765 79
BC026858.1 - 6 - 7 650 79

View Full Product Details