Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pus10
Entrez Gene ID
74467
Gene Name
pseudouridylate synthase 10
Gene Aliases
2810013G11Rik, 4933435A13Rik, AU014648, C77560, Ccdc139
Chromosome Location
Chr.11: 23665354 - 23732876 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033654.2 NP_001028826.1 4 - 5 541 60
NM_028304.2 NP_082580.1 4 - 5 954 60
NM_028956.4 NP_083232.1 4 - 5 551 60
GenBank mRNA AK012737.1 - 4 - 5 718 60
AK017063.1 - 4 - 5 528 60
AK028368.1 - 4 - 5 538 60
AK029866.1 - 4 - 5 542 60
AK088562.1 - 4 - 5 542 60
AK145055.1 - 4 - 5 900 60
BC025555.1 - 4 - 5 772 60

View Full Product Details