Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phf14
Entrez Gene ID
75725
Gene Name
PHD finger protein 14
Gene Aliases
1110001C23Rik, 4932409F11Rik, 5730446A07Rik, AA623952, AV297001, mKIAA0783
Chromosome Location
Chr.6: 11907599 - 12099859 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168382.1 NP_001161854.1 11 - 12 2467 81
NM_029404.3 NP_083680.2 11 - 12 2464 81
GenBank mRNA AK003209.1 - 4 - 5 553 81
AK016517.1 - 11 - 12 2467 81
AK173024.1 - 3 - 4 306 81
BC040236.1 - 11 - 12 2430 81

View Full Product Details